Anti-IgG2a Goat Polyclonal Antibody (CF™ 770)

Supplier: Biotium

20264 20264-1
71004-336EA 480.16 CAD
71004-336 71004-338
Anti-IgG2a Goat Polyclonal Antibody (CF™ 770)
Antibodies
Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: CF™ 770
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR