Anti-IgG Donkey Polyclonal Antibody (CF™ 770)

Supplier: Biotium

20279 20279-1
71004-348EA 411.57 CAD
71004-348 71004-350
Anti-IgG Donkey Polyclonal Antibody (CF™ 770)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 770
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR