Anti-IgG Donkey Polyclonal Antibody (CF™ 750)

Supplier: Biotium

20298 20298-1
71003-846EA 411.57 CAD
71003-846 71003-848
Anti-IgG Donkey Polyclonal Antibody (CF™ 750)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 750
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR