Anti-IgG Goat Polyclonal Antibody (CF™ 750)

Supplier: Biotium

20073 20073-1
89138-530EA 375.63 CAD
89138-530 71004-180
Anti-IgG Goat Polyclonal Antibody (CF™ 750)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 750
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR