Anti-IgG2 beta Goat Polyclonal Antibody (CF™ 633)

Supplier: Biotium

20270 20270-1
71003-830EA 440.99 CAD
71003-830 71003-832
Anti-IgG2 beta Goat Polyclonal Antibody (CF™ 633)
Antibodies
Type: Secondary
Antigen: IgG2 beta
Clonality: Polyclonal
Clone:
Conjugation: CF™ 633
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR