Anti-IgG Rabbit Polyclonal Antibody (CF™ 633)

Supplier: Biotium

20136 20136-1
89138-684EA 411.57 CAD
89138-684 89138-686
Anti-IgG Rabbit Polyclonal Antibody (CF™ 633)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 633
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR