Anti-IgG Goat Polyclonal Antibody (CF™ 633)

Supplier: Biotium

20133 20133-1
89138-672EA 411.57 CAD
89138-672 89138-674
Anti-IgG Goat Polyclonal Antibody (CF™ 633)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 633
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR