Anti-IgY Goat Polyclonal Antibody (CF™ 633)

Supplier: Biotium

20126 20126-1
89138-652EA 411.57 CAD
89138-652 89138-654
Anti-IgY Goat Polyclonal Antibody (CF™ 633)
Antibodies
Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: CF™ 633
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR