Anti-IgG Rabbit Polyclonal Antibody (CF™ 594)

Supplier: Biotium

20173 20173-1
97065-540EA 411.57 CAD
97065-540 97065-542
Anti-IgG Rabbit Polyclonal Antibody (CF™ 594)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 594
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR