Anti-IgG Goat Polyclonal Antibody (CF™ 594)

Supplier: Biotium

20118 20118-1
89138-628EA 342.99 CAD
89138-628 71004-278
Anti-IgG Goat Polyclonal Antibody (CF™ 594)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 594
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR