Anti-IgG Donkey Polyclonal Antibody (CF™ 568)

Supplier: Biotium

20377-500UL 20377-50UL
89427-286EA 411.57 CAD
89427-286 89427-288
Anti-IgG Donkey Polyclonal Antibody (CF™ 568)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 568
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Guinea Pig
Order Now

SPECIFICATIONS

Antigen symbol IgG
Environmentally Preferable
Antigen name Immunoglobulin G
Antibody type Secondary
Clonality Polyclonal
Conjugation CF® 568
Reactivity Guinea pig
Host Donkey
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Epitope Heavy and Light chain
Cross adsorption Minimal Cross-Reactivity: Bv, Ch, Gt, Hs, Hu, Ms, Rb, Sh, SHm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR