Anti-IgG Goat Polyclonal Antibody (CF™ 568)

Supplier: Biotium

20109 20109-1
89138-600EA 272.76 CAD
89138-600 10018-414
Anti-IgG Goat Polyclonal Antibody (CF™ 568)
Antibodies
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-mouse IgG (H+L) with CF™568 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Red fluorescent CF™568 (Ex/Em 562/583 nm) produces much brighter antibody conjugates than Alexa Fluor® 568, and has superior photostability.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 568
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR