Anti-IgG Rabbit Polyclonal Antibody (CF™ 568)

Supplier: Biotium

20093 20093-1
89138-550EA 411.57 CAD
89138-550 89138-552
Anti-IgG Rabbit Polyclonal Antibody (CF™ 568)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 568
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR