Anti-IgG Donkey Polyclonal Antibody (CF™ 568)

Supplier: Biotium

20092 20092-1
89138-546EA 522.35 CAD
89138-546 89138-548
Anti-IgG Donkey Polyclonal Antibody (CF™ 568)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 568
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR