Anti-IgG Goat Polyclonal Antibody (CF™ 555)

Supplier: Biotium

20232 20232-1
89138-788EA 458.29 CAD
89138-788 89138-790
Anti-IgG Goat Polyclonal Antibody (CF™ 555)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 555
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR