Anti-IgG Donkey Polyclonal Antibody (CF™ 555)

Supplier: Biotium

20038 20038-1
89138-460EA 518.16 CAD
89138-460 89138-462
Anti-IgG Donkey Polyclonal Antibody (CF™ 555)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 555
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR