Anti-IgG Rabbit Polyclonal Antibody (CF™ 488A)

Supplier: Biotium

20071 20071-1
71003-838EA 411.57 CAD
71003-838 71003-840
Anti-IgG Rabbit Polyclonal Antibody (CF™ 488A)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 488A
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR