Anti-IgY Rabbit Polyclonal Antibody (CF™ 488A)

Supplier: Biotium

20079 20079-1
71003-964EA 342.99 CAD
71003-964 71003-966
Anti-IgY Rabbit Polyclonal Antibody (CF™ 488A)
Antibodies
Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: CF™ 488A
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR