Anti-IgG2 beta Goat Polyclonal Antibody (CF™ 488A)

Supplier: Biotium

20266 20266-1
71004-034EA 440.99 CAD
71004-034 71004-036
Anti-IgG2 beta Goat Polyclonal Antibody (CF™ 488A)
Antibodies
Type: Secondary
Antigen: IgG2 beta
Clonality: Polyclonal
Clone:
Conjugation: CF™ 488A
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR