Anti-IgY Goat Polyclonal Antibody (CF™ 488A)

Supplier: Biotium

20020 20020-1
89138-412EA 411.57 CAD
89138-412 89138-414
Anti-IgY Goat Polyclonal Antibody (CF™ 488A)
Antibodies
Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: CF™ 488A
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR