Anti-IgG Goat Polyclonal Antibody (CF™ 405M)

Supplier: Biotium

20373-500UL 20373-50UL
89427-270EA 411.57 CAD
89427-270 89427-272
Anti-IgG Goat Polyclonal Antibody (CF™ 405M)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 405M
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR