Anti-IgG Goat Polyclonal Antibody (CF™ 405M)

Supplier: Biotium

20340-500UL 20340-50UL
89427-190EA 534.09 CAD
89427-190 89427-192
Anti-IgG Goat Polyclonal Antibody (CF™ 405M)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 405M
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR