Anti-IgG Chicken Polyclonal Antibody (CF™ 350)

Supplier: Biotium

20331-500UL 20331-50UL
89427-154EA 342.99 CAD
89427-154 89427-156
Anti-IgG Chicken Polyclonal Antibody (CF™ 350)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 350
Epitope: Heavy and Light chain
Host: Chicken
Isotype: IgY
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR