Anti-IgG2a Goat Polyclonal Antibody (CF™ 350)

Supplier: Biotium

20255 20255-1
71004-062EA 440.99 CAD
71004-062 71004-064
Anti-IgG2a Goat Polyclonal Antibody (CF™ 350)
Antibodies
Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: CF™ 350
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR