Anti-IgG Goat Polyclonal Antibody (CF™ 350)

Supplier: Biotium

20140 20140-1
89138-694EA 342.99 CAD
89138-694 71004-298
Anti-IgG Goat Polyclonal Antibody (CF™ 350)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 350
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR