Anti-IgG Isotype Ctrl Armenian Hamster Monoclonal Antibody [clone: HTK888]

Supplier: Biolegend
400901 400902
CA400901-BLEA 75 CAD
CA400901-BL CA400902-BL
Anti-IgG Isotype Ctrl Armenian Hamster Monoclonal Antibody [clone: HTK888]
Antibodies
Purified Armenian Hamster IgG Isotype Ctrl [HTK888]; Isotype: Armenian Hamster IgG; Reactivity: Armenian Hamster; Apps: FC, ICFC, ICC, IF, IP, WB; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control, and the purity is greater than 95% by SDS-PAGE. Use at concentrations comparable to those of the specific antibody of interest.

Type: Control
Antigen: IgG Isotype Ctrl
Clonality: Monoclonal
Clone: HTK888
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Armenian Hamster
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR