Anti-XBP-1s Mouse Monoclonal Antibody [clone: 143F]

Supplier: Biolegend
647501 647502
CA647501-BLEA 197.88 CAD
CA647501-BL CA647502-BL
Anti-XBP-1s Mouse Monoclonal Antibody [clone: 143F]
Antibodies
Purified anti-XBP-1s [143F]; Isotype: Mouse IgG2a; Reactivity: Human; Apps: WB, IF, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 1 to 2 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: XBP-1s
Clonality: Monoclonal
Clone: 143F
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR