Anti-TIF1b Mouse Monoclonal Antibody [clone: 20A1]

Supplier: Biolegend
619301 619302
CA619301-BLEA 161.18 CAD
CA619301-BL CA619302-BL
Anti-TIF1b Mouse Monoclonal Antibody [clone: 20A1]
Antibodies
Purified anti-TIF1β (KAP-1, TRIM28) [20A1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Mouse; Apps: WB, IF, ChIP; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 5 μg per 5 ml antibody dilution buffer for each mini-gel. For immunofluorescence microscopy, a concentration range of 2.5-10 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TIF1b
Clonality: Monoclonal
Clone: 20A1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR