Anti-CD90.1 Mouse Monoclonal Antibody [clone: OX-7]

Supplier: Biolegend
202501 202502
CA202501-BLEA 197.88 CAD
CA202501-BL CA202502-BL
Anti-CD90.1 Mouse Monoclonal Antibody [clone: OX-7]
Antibodies
Purified anti-rat CD90/mouse CD90.1 (Thy-1.1) [OX-7]; Isotype: Mouse IgG1, κ; Reactivity: Rat, Mouse (AKR/J and PL mouse strains). Cross-Reactivity: Rabbit (Lapine), Guinea Pig6; Apps: FC, IF, IHC, IP, WB; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD90.1
Clonality: Monoclonal
Clone: OX-7
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Rat;Mouse (AKR/J and PL mouse strains). Cross-Reactivity: Rabbit (Lapine);Guinea Pig6
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR