Anti-CD81 Armenian Hamster Monoclonal Antibody [clone: Eat-2]

Supplier: Biolegend
104901 104902
CA104901-BLEA 132.45 CAD
CA104901-BL CA104902-BL
Anti-CD81 Armenian Hamster Monoclonal Antibody [clone: Eat-2]
Antibodies
Purified anti-mouse/rat CD81 [Eat-2]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Rat; Apps: FC, IP, WB; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD81
Clonality: Monoclonal
Clone: Eat-2
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse;Rat
Order Now

SPECIFICATIONS

Antigen symbol CD81
Environmentally Preferable
Antigen synonyms TAPA-1
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone Eat-2
Reactivity Rat,Mouse
Host Armenian hamster
Gene ID 12520, 25621
Isotype IgG
Western blot Yes
ImmunoPrecipitation Yes
Flow cytometry Yes
Immunogen CD81+ mouse B lymphoma 38C13
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Purification The antibody was purified by affinity chromatography.
Concentration 0.50 mg/mL
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C.
Shipping temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR