Anti-Mac-2 (Galectin-3) Rat Monoclonal Antibody [clone: M3/38]

Supplier: Biolegend
125401 125402
CA125401-BLEA 114.9 CAD
CA125401-BL CA125402-BL
Anti-Mac-2 (Galectin-3) Rat Monoclonal Antibody [clone: M3/38]
Antibodies
Purified anti-mouse/human Mac-2 (Galectin-3) [M3/38]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, Human; Apps: FC, IF, WB, IP, IHC, ELISA; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Mac-2 (Galectin-3)
Clonality: Monoclonal
Clone: M3/38
Conjugation:
Epitope:
Host: Rat
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR