Anti-IL-10 Rat Monoclonal Antibody [clone: JES5-16E3]

Supplier: Biolegend
505001 505002
CA505001-BLEA 132.45 CAD
CA505001-BL CA505002-BL
Anti-IL-10 Rat Monoclonal Antibody [clone: JES5-16E3]
Antibodies
Purified anti-mouse IL-10 [JES5-16E3]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: ELISA Capture, CyTOF®, IHC; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture applications, a concentration range of 0.1-1.0 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application. To obtain a linear standard curve, serial dilutions of IL-10 recombinant protein ranging from 2000 to 15 pg/ml are recommended for each ELISA plate. For ELISA capture, Carbonate buffer, pH 9.5 is highly recommended for coating buffer.

Type: Primary
Antigen: IL-10
Clonality: Monoclonal
Clone: JES5-16E3
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR