Anti-IFN-g Rat Monoclonal Antibody [clone: AN-18]

Supplier: Biolegend
517901 517902
CA517901-BLEA 142.03 CAD
CA517901-BL CA517902-BL
Anti-IFN-g Rat Monoclonal Antibody [clone: AN-18]
Antibodies
Purified anti-mouse IFN-γ [AN-18]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ELISA Capture, ELISPOT, IP; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture applications, a concentration range of 0.25-2.0 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of IFN-γ recombinant protein ranging from 3.9 to 500 pg/mL are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IFN-g
Clonality: Monoclonal
Clone: AN-18
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR