Anti-F4/80 Rat Monoclonal Antibody [clone: BM8]

Supplier: Biolegend
123101 123102
CA123101-BLEA 75 CAD
CA123101-BL CA123102-BL
Anti-F4/80 Rat Monoclonal Antibody [clone: BM8]
Antibodies
Purified anti-mouse F4/80 [BM8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC, WB; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: F4/80
Clonality: Monoclonal
Clone: BM8
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR