Anti-CD90 Rat Monoclonal Antibody [clone: G7]

Supplier: Biolegend
105201 105202
CA105201-BLEA 86.17 CAD
CA105201-BL CA105202-BL
Anti-CD90 Rat Monoclonal Antibody [clone: G7]
Antibodies
Purified anti-mouse CD90 [G7]; Isotype: Rat IgG2c, κ; Reactivity: Mouse; Apps: FC, IHC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD90
Clonality: Monoclonal
Clone: G7
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2c, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR