Anti-CD8a Rat Monoclonal Antibody [clone: 5H10-1]

Supplier: Biolegend
100801 100802
CA100801-BLEA 132.45 CAD
CA100801-BL CA100802-BL
Anti-CD8a Rat Monoclonal Antibody [clone: 5H10-1]
Antibodies
Purified anti-mouse CD8a [5H10-1]; Isotype: Rat (SD) IgG2b, λ; Reactivity: Mouse; Apps: FC, IF, IHC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: 5H10-1
Conjugation:
Epitope:
Host: Rat
Isotype: Rat (SD) IgG2b, λ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR