Anti-CD28 Syrian Hamster Monoclonal Antibody [clone: 37.51]

Supplier: Biolegend
102101 102102
CA102101-BLEA 86.17 CAD
CA102101-BL CA102102-BL
Anti-CD28 Syrian Hamster Monoclonal Antibody [clone: 37.51]
Antibodies
Purified anti-mouse CD28 [37.51]; Isotype: Syrian Hamster IgG; Reactivity: Mouse; Apps: FC, IHC, IP; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: 37.51
Conjugation:
Epitope:
Host: Syrian Hamster
Isotype: Syrian Hamster IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR