Anti-CD279 (PD-1) Rat Monoclonal Antibody [clone: RMP1-14]

Supplier: Biolegend
114101 114102
CA114101-BLEA 132.45 CAD
CA114101-BL CA114102-BL
Anti-CD279 (PD-1) Rat Monoclonal Antibody [clone: RMP1-14]
Antibodies
Purified anti-mouse CD279 (PD-1) [RMP1-14]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC, WB; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD279 (PD-1)
Clonality: Monoclonal
Clone: RMP1-14
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR