Anti-CD18 Rat Monoclonal Antibody [clone: M18/2]

Supplier: Biolegend
101401 101402
CA101401-BLEA 86.17 CAD
CA101401-BL CA101402-BL
Anti-CD18 Rat Monoclonal Antibody [clone: M18/2]
Antibodies
Purified anti-mouse CD18 [M18/2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC, IP, WB; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD18
Clonality: Monoclonal
Clone: M18/2
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

SPECIFICATIONS

Antigen symbol CD18
Environmentally Preferable
Antigen synonyms Integrin β2 subunit|ITGB2|b2 integrin
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone M18/2
Reactivity Mouse
Host Rat
Gene ID 16414
Isotype IgG2a kappa
Western blot Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
Flow cytometry Yes
Immunogen Mouse CTL glycoprotein
Form Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
Purification The antibody was purified by affinity chromatography.
Concentration 0.50 mg/mL
Storage temperature The antibody solution should be stored undiluted between 2 °C and 8 °C.
Shipping temperature Ambient RT

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR