Anti-IL-8 Mouse Monoclonal Antibody [clone: BH0814]

Supplier: Biolegend
514601 514602
CA514601-BLEA 169.15 CAD
CA514601-BL CA514602-BL
Anti-IL-8 Mouse Monoclonal Antibody [clone: BH0814]
Antibodies
Purified anti-human IL-8 [BH0814]; Isotype: Mouse IgG2b, λ; Reactivity: Human; Apps: ELISA Capture; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture applications, a concentration range of 2-6 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of IL-8 recombinant protein ranging from 2000 to 15 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-8
Clonality: Monoclonal
Clone: BH0814
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, λ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR