Anti-IL-12 (p70) Rat Monoclonal Antibody [clone: 7B12]

Supplier: Biolegend
511001 511002
CA511001-BLEA 169.15 CAD
CA511001-BL CA511002-BL
Anti-IL-12 (p70) Rat Monoclonal Antibody [clone: 7B12]
Antibodies
Purified anti-human IL-12 (p70) [7B12]; Isotype: Rat IgG1; Reactivity: Human, p70 heterodimer; Apps: ELISA Capture; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture applications, a concentration range of 1-4 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of IL-12 p70 recombinant protein ranging from 1000 to 8 pg/ml are recommended for each ELISA plate. For use as an ELISPOT capture antibody, a concentration range of 2-6 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-12 (p70)
Clonality: Monoclonal
Clone: 7B12
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG1
Reactivity: Human;p70 heterodimer
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR