Anti-IL-10 Rat Monoclonal Antibody [clone: JES3-19F1]

Supplier: Biolegend
506801 506802
CA506801-BLEA 132.45 CAD
CA506801-BL CA506802-BL
Anti-IL-10 Rat Monoclonal Antibody [clone: JES3-19F1]
Antibodies
Purified anti-human IL-10 [JES3-19F1]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: ELISA Capture, IHC, WB; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture applications, a concentration range of 2-6 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of IL-10 recombinant protein ranging from 250 to 2 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-10
Clonality: Monoclonal
Clone: JES3-19F1
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR