Anti-IL-1RA Mouse Monoclonal Antibody [clone: JK1RA-1]

Supplier: Biolegend
509901 509902
CA509901-BLEA 169.15 CAD
CA509901-BL CA509902-BL
Anti-IL-1RA Mouse Monoclonal Antibody [clone: JK1RA-1]
Antibodies
Purified anti-human IL-1 RA [JK1RA-1]; Isotype: Mouse IgG1; Reactivity: Human; Apps: ELISA Capture; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture applications, a concentration range of 4-6 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-1RA
Clonality: Monoclonal
Clone: JK1RA-1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR