Anti-IFN-g Mouse Monoclonal Antibody [clone: MD-1]

Supplier: Biolegend
507501 507502
CA507501-BLEA 169.15 CAD
CA507501-BL CA507502-BL
Anti-IFN-g Mouse Monoclonal Antibody [clone: MD-1]
Antibodies
Purified anti-human IFN-γ [MD-1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus; Apps: ELISA Capture, ICFC, IHC, WB; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA Capture applications, the antibody should be titrated between 0.25 - 2 μg/ml to determine optimal condition. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IFN-g
Clonality: Monoclonal
Clone: MD-1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR