Anti-G-CSF Rat Monoclonal Antibody [clone: BVD13-3A5]

Supplier: Biolegend
502101 502102
CA502101-BLEA 150.01 CAD
CA502101-BL CA502102-BL
Anti-G-CSF Rat Monoclonal Antibody [clone: BVD13-3A5]
Antibodies
Purified anti-human G-CSF [BVD13-3A5]; Isotype: Rat IgG1, κ; Reactivity: Human; Apps: ELISA Capture, IHC; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture applications, a concentration range of 1-4 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: G-CSF
Clonality: Monoclonal
Clone: BVD13-3A5
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR