Anti-CD55 Mouse Monoclonal Antibody [clone: JS11]

Supplier: Biolegend
311301 311302
CA311301-BLEA 103.73 CAD
CA311301-BL CA311302-BL
Anti-CD55 Mouse Monoclonal Antibody [clone: JS11]
Antibodies
Purified anti-human CD55 [JS11]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD55
Clonality: Monoclonal
Clone: JS11
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR