Anti-CD324 (E-Cadherin) Mouse Monoclonal Antibody [clone: 67A4]

Supplier: Biolegend
324101 324102
CA324101-BLEA 161.18 CAD
CA324101-BL CA324102-BL
Anti-CD324 (E-Cadherin) Mouse Monoclonal Antibody [clone: 67A4]
Antibodies
Purified anti-human CD324 (E-Cadherin) [67A4]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, IF, WB; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per million cells in 100 μl volume. For Western blotting, the suggested use of this reagent is 0.1 - 1.0 μg per ml (dilution 1:5000 - 1:500). It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD324 (E-Cadherin)
Clonality: Monoclonal
Clone: 67A4
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR