Anti-CD117 Mouse Monoclonal Antibody [clone: 104D2]

Supplier: Biolegend
313201 313202
CA313201-BLEA 122.88 CAD
CA313201-BL CA313202-BL
Anti-CD117 Mouse Monoclonal Antibody [clone: 104D2]
Antibodies
Purified anti-human CD117 (c-kit) [104D2]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Cattle (Bovine, Cow); Apps: FC, CyTOF®, IF, IP; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 2.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD117
Clonality: Monoclonal
Clone: 104D2
Conjugation:
Epitope:
Host: Mouse
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR