Anti-DNA-PKcs Phospho (Thr2609) Mouse Monoclonal Antibody [clone: 10B1]

Supplier: Biolegend
612901 612902
CA612901-BLEA 188.3 CAD
CA612901-BL CA612902-BL
Anti-DNA-PKcs Phospho (Thr2609) Mouse Monoclonal Antibody [clone: 10B1]
Antibodies
Purified anti-DNA-PKcs Phospho (Thr2609) [10B1]; Isotype: Mouse IgG1, κ; Reactivity: Human, reacts with Thr2609-phosphorylated DNA-PKcs; Apps: IF; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescence staining. For immunofluorescence microscopy, a concentration range of 5 - 10 μg per ml is suggested. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: DNA-PKcs Phospho (Thr2609)
Clonality: Monoclonal
Clone: 10B1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;reacts with Thr2609-phosphorylated DNA-PKcs
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR