Anti-Cytokeratin 18 Mouse Monoclonal Antibody [clone: DA-7]

Supplier: Biolegend
628401 628402
CA628401-BLEA 161.18 CAD
CA628401-BL CA628402-BL
Anti-Cytokeratin 18 Mouse Monoclonal Antibody [clone: DA-7]
Antibodies
Purified anti-Cytokeratin 18 [DA-7]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, IF, ELISA, ICC, IHC, IP; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 5 μg antibody per 5 ml antibody dilution buffer for each mini-gel. For immunofluorescence microscopy: Use a dilution range of 1-4 μg/ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Cytokeratin 18
Clonality: Monoclonal
Clone: DA-7
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR